Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Meat Peptone

Catalog No. AAH2669422
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging Type:
Glass Bottle
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
AAH2669422 100 g Plastic Bottle
AAH2669436 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2669422 Supplier Thermo Scientific Chemicals Supplier No. H2669422
Add to Cart

Peptone, from meat (pork gelatine), CAS # 91079-38-8, is a nutrient isolated from meat used to promote the in vitro growth of several species including bacteria and fungi.

Peptone is used in nutrient media for growing bacteria and fungiMeat Peptone is used for routine and mass scale cultivation of organisms, which is utilized for the production of antibiotics, enzymes and vitamin production. It is also useful for the cultivation of a variety of microorganisms. Further, it is used in culture media, industry fermentation and the pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Peptone, from meat, is a growth-promoting nutrient used in bacteria and fungi cultures

Applications

  • It is commonly used in animal cell culture.
  • Peptone is used in mass scale organism cultivation, and fermentation processes
  • Peptone, from meat, is used as a supplement to in vitro culture media to help to increase cell number, specific monoclonal antibody production, and maintain long-term cultures
  • This compound has been used as a component in Ax-2 axenic medium to induce reproduction of Dictyostelium discoideum
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Packaging Type Plastic Bottle
Quantity 100 g
Sensitivity Hygroscopic

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.