Learn More
PCE AMERICAS INC Handheld pH meat Meter 0.00 to 14.00 pH
SDP
Catalog No. NC3654417
Encompass_Preferred
"The food pH meter PCE-228M is a very easy to use hand-held pH meter for food. At the food pH Meter can the pH value and the temperature be read off via the large LCD display or saved to the SD card (Excel file). Another option is the online transmission via the RS-232 interface directly from the food pH meter to a PC or laptop. For this, we offer optional software and the appropriate RS-232 interface cable as an accessory.The temperature compensation is carried out either manually or automatically via the supplied stainless steel temperature sensor. All this allows a fast and reliable pH measurement of all foods, e.g. sausage, meat and cheese. The food pH meter has a three-point calibration, which is performed automatically. The pH meter is battery operated (6 x 1.5 V batteries are included). The pH-Meter PCE-228M is ideal for occasional pH measurements in the laboratory."
Catalog No. NC3654417 Supplier PCE AMERICAS INC Supplier No. PCE228M
"The food pH meter PCE-228M is a very easy to use hand-held pH meter for food. At the food pH Meter can the pH value and the temperature be read off via the large LCD display or saved to the SD card (Excel file). Another option is the online transmission via the RS-232 interface directly from the food pH meter to a PC or laptop. For this, we offer optional software and the appropriate RS-232 interface cable as an accessory.The temperature compensation is carried out either manually or automatically via the supplied stainless steel temperature sensor. All this allows a fast and reliable pH measurement of all foods, e.g. sausage, meat and cheese. The food pH meter has a three-point calibration, which is performed automatically. The pH meter is battery operated (6 x 1.5 V batteries are included). The pH-Meter PCE-228M is ideal for occasional pH measurements in the laboratory."