Learn More
Selleck Chemical LLC Mebendazole-100mg
SDP
Catalog No. 502390054
Encompass_Preferred
Mebendazole (Vermox, Telmin, Pantelmin, Mebenvet) is a synthetic benzimidazole derivate and anthelmintic agent. Mebendazole interferes with the reproduction and survival of helminths by inhibiting the formation of their cytoplasmic microtubules, thereby selectively and irreversibly blocking glucose uptake.
Catalog No. 50-239-0054 Supplier Selleck Chemical LLC Supplier No. S4610100MG
Mebendazole (Vermox, Telmin, Pantelmin, Mebenvet) is a synthetic benzimidazole derivate and anthelmintic agent. Mebendazole interferes with the reproduction and survival of helminths by inhibiting the formation of their cytoplasmic microtubules, thereby selectively and irreversibly blocking glucose uptake.