Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mebendazole-amine 5mg | 52329-60-9 | 237.26 g/mol | C14H11N3O | 5 MG
SDP

Catalog No. 5000228248
Encompass_Preferred

Mebendazole-amine is a research metabolite of mebendazole supplied as a high-purity solid for analytical and metabolic studies. Store protected from light and follow solvent storage recommendations for long-term stability.

  • Research metabolite of mebendazole.
  • High purity: 98.35%.
  • Molecular weight 237.26 g/mol; formula C14H11N3O.
  • Supplied in small vials suitable for analytical work (e.g., 5 mg).
  • Appearance: solid, white to off-white.
  • Storage: 4°C, protect from light; in solvent, -80°C up to 6 months or -20°C up to 1 month.

Catalog No. 50-002-28248 Supplier Medchemexpress LLC Supplier No. HY1147505MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mebendazole-amine is a research metabolite of mebendazole supplied as a high-purity solid for analytical and metabolic studies. Store protected from light and follow solvent storage recommendations for long-term stability.

  • Research metabolite of mebendazole.
  • High purity: 98.35%.
  • Molecular weight 237.26 g/mol; formula C14H11N3O.
  • Supplied in small vials suitable for analytical work (e.g., 5 mg).
  • Appearance: solid, white to off-white.
  • Storage: 4°C, protect from light; in solvent, -80°C up to 6 months or -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.