Learn More
Medchemexpress LLC Mebeverine acid-d5, hydrochloride | 475203-77-1 | 99.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY12769S11MG
Encompass_Preferred
Mebeverine acid-d5 hydrochloride is a deuterium labeled form of Mebeverine acid hydrochloride, a secondary metabolite of the intestinal antispasmodic agent Mebeverine. It is formed by the body's hydrolysis of Mebeverine, acting as a key circulating metabolite and an important marker for oral Mebeverine. This compound is intended for research use only, leveraging stable heavy isotopes for quantitation in drug development.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-40781