Learn More
Medchemexpress LLC Mebeverine acid-d5 hydrochloride | 475203-77-1 | 99.0% | 10 MG
SDP
Catalog No. 5000339750
Encompass_Preferred
Mebeverine acid-d5 hydrochloride is a deuterium-labeled form of Mebeverine acid hydrochloride, a secondary metabolite of the intestinal antispasmodic agent Mebeverine. This metabolite is formed in the body through hydrolysis and serves as a key circulating marker for oral Mebeverine.
  • Deuterium-labeled compound
  • Secondary metabolite of mebeverine
  • Key circulating metabolite for oral mebeverine
  • Used as a tracer and internal standard
  • Suitable for quantitative analysis methods
  • Purity of 99.0%
Catalog No. 50-003-39750 Supplier Medchemexpress LLC Supplier No. HY12769S110MG
Mebeverine acid-d5 hydrochloride is a deuterium-labeled form of Mebeverine acid hydrochloride, a secondary metabolite of the intestinal antispasmodic agent Mebeverine. This metabolite is formed in the body through hydrolysis and serves as a key circulating marker for oral Mebeverine.
  • Deuterium-labeled compound
  • Secondary metabolite of mebeverine
  • Key circulating metabolite for oral mebeverine
  • Used as a tracer and internal standard
  • Suitable for quantitative analysis methods
  • Purity of 99.0%