Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mebeverine alcohol | 14367-47-6 | 99.6% | 10 MG
SDP

Catalog No. 5000342427
Encompass_Preferred

Mebeverine alcohol is a metabolite of Mebeverine, a musculotropic antispasmodic drug. This product is provided as a liquid with a light yellow to yellow appearance and boasts a high purity of 99.61%.

  • Molecular weight: 265.39
  • Chemical formula: C16H27NO2
  • CAS number: 14367-47-6
  • Appearance: Liquid, light yellow to yellow
  • Purity: 99.61%
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Solubility in DMSO: 100 mg/mL

Catalog No. 50-003-42427 Supplier Medchemexpress LLC Supplier No. HY1277010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mebeverine alcohol is a metabolite of Mebeverine, a musculotropic antispasmodic drug. This product is provided as a liquid with a light yellow to yellow appearance and boasts a high purity of 99.61%.

  • Molecular weight: 265.39
  • Chemical formula: C16H27NO2
  • CAS number: 14367-47-6
  • Appearance: Liquid, light yellow to yellow
  • Purity: 99.61%
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Solubility in DMSO: 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.