Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mebeverine alcohol | 14367-47-6 | MFCD01656164 | 99.6% | 265.39 g/mol | C16H27NO2 | 5 MG
SDP

Catalog No. 5000263972
Encompass_Preferred

Mebeverine alcohol is the primary metabolite of mebeverine supplied as an analytical standard for laboratory research and analytical applications. It is intended for research use only and not for human administration.

  • Major metabolite of mebeverine.
  • Analytical standard for research and analytical applications.
  • High purity: 99.6%.
  • Chemical formula C16H27NO2; molecular weight 265.39 g/mol.
  • Available in small pack sizes (for example, 5 mg) suitable for analytical assays.
  • Storage recommendations: pure form stable at -20°C and 4°C; in solvent keep frozen for long-term storage.

Catalog No. 50-002-63972 Supplier Medchemexpress LLC Supplier No. HY127705MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mebeverine alcohol is the primary metabolite of mebeverine supplied as an analytical standard for laboratory research and analytical applications. It is intended for research use only and not for human administration.

  • Major metabolite of mebeverine.
  • Analytical standard for research and analytical applications.
  • High purity: 99.6%.
  • Chemical formula C16H27NO2; molecular weight 265.39 g/mol.
  • Available in small pack sizes (for example, 5 mg) suitable for analytical assays.
  • Storage recommendations: pure form stable at -20°C and 4°C; in solvent keep frozen for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.