Learn More
Medchemexpress LLC Mebeverine hydrochloride | 2753-45-9 | 99.9% | C25H36ClNO5 | 25 MG
SDP
Catalog No. 5000357974
Encompass_Preferred
Mebeverine hydrochloride is a β-phenylethylamine derivative and a musculotropic agent that potently blocks intestinal peristalsis. It directly blocks voltage-operated sodium channels and inhibits intracellular calcium accumulation.
  • Potent blocker of intestinal peristalsis
  • Directly blocks voltage-operated sodium channels
  • Inhibits intracellular calcium accumulation
  • Relevant for metabolic or endocrine disease research
  • Targets sodium channels
Catalog No. 50-003-57974 Supplier Medchemexpress LLC Supplier No. 25MGHYA0078
Mebeverine hydrochloride is a β-phenylethylamine derivative and a musculotropic agent that potently blocks intestinal peristalsis. It directly blocks voltage-operated sodium channels and inhibits intracellular calcium accumulation.
  • Potent blocker of intestinal peristalsis
  • Directly blocks voltage-operated sodium channels
  • Inhibits intracellular calcium accumulation
  • Relevant for metabolic or endocrine disease research
  • Targets sodium channels