Learn More
Selleck Chemical LLC Mebeverine Hydrochloride S5027-25mg
SDP
Catalog No. 503111960
Encompass_Preferred
Mebeverine (Duspatalin Duspatal Colofac) a 3-phenylethylamine derivative of methoxybenzamine acts as a musculotropic antispasmodic agent
Catalog No. 50-311-1960 Supplier Selleck Chemical LLC Supplier No. S502725MG
Mebeverine (Duspatalin Duspatal Colofac) a 3-phenylethylamine derivative of methoxybenzamine acts as a musculotropic antispasmodic agent