Learn More
Advanced Biomatrix Mebiol Thermoreversible Hydrogel, 10mL
Catalog No. NC1409870
Encompass_Preferred
Mebiol is a thermoreversible hydrogel that gels at 37C and transitions back to a liquid at 4C. Mebiol is also biologically inert, allowing for easy spheroid formation in the hydrogel phase, and easy cell retrieval during the cold-melt phase. Mebiol Thermoreversible Hydrogel is 1 gram of a copolymer of poly(N-isopropylacrylamide) and poly(ethylene glycol) (PNIPAAm-PEG). The defining feature of Mebiol is its temperature reversible sol-gel transition. Chill Mebiol to turn it into a sol (meaning it handles like a liquid). Heat Mebiol to turn it into a solid hydrogel. The unique thermoreversible feature allows for extremely easy cell handling. Mebiol is easy to use, non-toxic, 100% synthetic and has high transparency. Published applications include stem cell and pluripotent stem cell culture, expansion, and differentiation, spheroid culture, cell implantation, organ and tissue regeneration, drug delivery and more.
Catalog No. NC1409870 Supplier Advanced Biomatrix Supplier No. 518010ML
Mebiol is a thermoreversible hydrogel that gels at 37C and transitions back to a liquid at 4C. Mebiol is also biologically inert, allowing for easy spheroid formation in the hydrogel phase, and easy cell retrieval during the cold-melt phase. Mebiol Thermoreversible Hydrogel is 1 gram of a copolymer of poly(N-isopropylacrylamide) and poly(ethylene glycol) (PNIPAAm-PEG). The defining feature of Mebiol is its temperature reversible sol-gel transition. Chill Mebiol to turn it into a sol (meaning it handles like a liquid). Heat Mebiol to turn it into a solid hydrogel. The unique thermoreversible feature allows for extremely easy cell handling. Mebiol is easy to use, non-toxic, 100% synthetic and has high transparency. Published applications include stem cell and pluripotent stem cell culture, expansion, and differentiation, spheroid culture, cell implantation, organ and tissue regeneration, drug delivery and more.