Learn More
Lonza Walkersville MEBM Mammary Epithelial Cell Growth Basal Medium
Catalog No. NC9586599
Encompass
MEBM is a mammary epithelial cell basal medium sold separately. This can be used with 1 kit of CC-4136 (mammary epithelial cell growth medium SingleQuots Supplements and Growth Factors). The full MEGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3150.MEGM Mammary Epithelial Cell Growth Medium BulleKit has been optimized for the growth of Mammary Epithelial cells in a serum-free environment. This media system has been referenced in over 130 unique publications primarily focusing on breast cancer research. Content:1 x Bottle Basal Medium 500 mL
Catalog No. NC9586599 Supplier Lonza Walkersville Supplier No. CC3151
MEBM is a mammary epithelial cell basal medium sold separately. This can be used with 1 kit of CC-4136 (mammary epithelial cell growth medium SingleQuots Supplements and Growth Factors). The full MEGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3150.MEGM Mammary Epithelial Cell Growth Medium BulleKit has been optimized for the growth of Mammary Epithelial cells in a serum-free environment. This media system has been referenced in over 130 unique publications primarily focusing on breast cancer research. Content:1 x Bottle Basal Medium 500 mL