Learn More
Lonza Walkersville MEBM Mammary Epithelial Cell Growth Basal Medium, Phenol Red Free
Catalog No. NC9596813
Encompass
MEBM is a phenol red-free mammary epithelial cell basal medium sold separately. This can be used with 1 kit of CC-4136 (mammary epithelial cell growth medium SingleQuots Supplements and Growth Factors).Mammary Epithelial Cell Growth Medium has been optimized for the growth of Mammary Epithelial cells in a serum-free environment. This media system has been referenced in over 130 unique publications primarily focusing on breast cancer research. Content:1 x Bottle Basal Medium 500 mL
Catalog No. NC9596813 Supplier Lonza Walkersville Supplier No. CC3153
MEBM is a phenol red-free mammary epithelial cell basal medium sold separately. This can be used with 1 kit of CC-4136 (mammary epithelial cell growth medium SingleQuots Supplements and Growth Factors).Mammary Epithelial Cell Growth Medium has been optimized for the growth of Mammary Epithelial cells in a serum-free environment. This media system has been referenced in over 130 unique publications primarily focusing on breast cancer research. Content:1 x Bottle Basal Medium 500 mL