Learn More
Medchemexpress LLC HY-B1684 10mg Medchemexpress, Mebrofenin CAS:78266-06-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB168410MG
Encompass_Preferred
Medchemexpress, HY-B1684 10mg Mebrofenin CAS:78266-06-5 Mebrofenin (SQ 26962) is a type of iminodiacetic acid (IDA). Mebrofenin is available as a ready to use the kit for radio-labeling with Tc-99m. Tc-99m Mebrofenin, a diagnostic agent, is used for hepatobiliary imaging. Tc-99m Mebrofenin is the radiopharmaceutical of choice for the evaluation of hepatic function. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0156