Learn More
Medchemexpress LLC HY-P7250 10ug Medchemexpress, MEC/CCL28, Human CAS: Purity:>98%
SDP
Catalog No. 502083135
Encompass_Preferred
Medchemexpress, HY-P7250 10ug MEC/CCL28, Human CAS: Recombinant Human MEC/CCL28 is a CC chemokine, which plays a key role in the segregation and the compartmentalization of the mucosal immune system through recruitment of immune cells to specific locations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3135 Supplier Medchemexpress LLC Supplier No. HYP725010UG
Medchemexpress, HY-P7250 10ug MEC/CCL28, Human CAS: Recombinant Human MEC/CCL28 is a CC chemokine, which plays a key role in the segregation and the compartmentalization of the mucosal immune system through recruitment of immune cells to specific locations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.