Learn More
Medchemexpress LLC MEC/CCL28 Mouse 10ug
SDP
Catalog No. 5000181582
Encompass_Preferred
MEC/CCL28 Protein Mouse is a CC chemokine that is present in almost all mucosal tissues and acts as a unifying immunostimulant on mucosal surfaces CCL28 can bind to CCR3 and CCR10 to mediate immune responses viral infections cancer and antimicrobial effects MEC/CCL28 Protein Mouse is a recombinant mouse MEC/CCL28 (S20-R130) protein expressed by E coli[1][2]
Catalog No. 50-001-81582 Supplier Medchemexpress LLC Supplier No. HYP725110UG
MEC/CCL28 Protein Mouse is a CC chemokine that is present in almost all mucosal tissues and acts as a unifying immunostimulant on mucosal surfaces CCL28 can bind to CCR3 and CCR10 to mediate immune responses viral infections cancer and antimicrobial effects MEC/CCL28 Protein Mouse is a recombinant mouse MEC/CCL28 (S20-R130) protein expressed by E coli[1][2]