Learn More
Medchemexpress LLC HY-B1395 5mg Medchemexpress, Mecamylamine (hydrochloride) CAS:826-39-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB13955MG
Encompass_Preferred
Medchemexpress, HY-B1395 5mg Mecamylamine (HCl) CAS:826-39-1 Mecamylamine hydrochloride is an orally active, nonselective, noncompetitive nAChR antagonist that can treat various neuropsychiatric disorders. Mecamylamine hydrochloride is originally used as a ganglionic blocker in treating hypertension. Mecamylamine hydrochloride can easily crosses the blood-brain barrier. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0150