Learn More
Selleck Chemical LLC Mecamylamine Hydrochloride S9459-5MG
SDP
Catalog No. 5000024621
Encompass_Preferred
Mecamylamine hydrochloride is an orally active nonselective noncompetitive nAChR(nicotinic acetylcholine receptors) antagonist It is also a ganglionic blocker and can across the blood-brain barrier It can be used in the research of neuropsychiatric disorders hypertension antidepressant area
Catalog No. 50-000-24621 Supplier Selleck Chemical LLC Supplier No. S94595MG
Mecamylamine hydrochloride is an orally active nonselective noncompetitive nAChR(nicotinic acetylcholine receptors) antagonist It is also a ganglionic blocker and can across the blood-brain barrier It can be used in the research of neuropsychiatric disorders hypertension antidepressant area