Learn More
Cayman Chemical MecamylamInhydrochlorIde 50mg
SDP
Catalog No. 502971491
Encompass_Preferred
A noncompetitive nicotinic acetylcholine receptor antagonist with preferential activity at the α3β4 subtype (IC50 = 90-640 nM) compared to α4β2, α3β2, and α7 subtypes (IC50s range from 1-7 μM)
Catalog No. 50-297-1491 Supplier Cayman Chemical Supplier No. 1460250 MG
A noncompetitive nicotinic acetylcholine receptor antagonist with preferential activity at the α3β4 subtype (IC50 = 90-640 nM) compared to α4β2, α3β2, and α7 subtypes (IC50s range from 1-7 μM)