Learn More
Apexbio Technology LLC Mecarbinate 15574-49-9 10mg
SDP
Catalog No. 502240790
Encompass_Preferred
Mecarbinate (CAS 15574-49-9) is a small-molecule inhibitor targeting hepatitis C virus (HCV) replication It is designed to inhibit viral replication thereby interfering with essential HCV biological processes Mecarbinate exerts its biological activity primarily through disruption of viral RNA replication potentially by inhibiting viral RNA polymerase or host factors necessary for genome replication In in vitro assays Mecarbinate demonstrates inhibition of HCV RNA replication efficiency with reported IC50 values varying according to experimental context and assay specifics Based on these pharmacological properties Mecarbinate holds research potential in studying viral infection mechanisms and aiding antiviral therapy screening
This item is not returnable. View return policy
Catalog No. 50-224-0790 Supplier Apexbio Technology LLC Supplier No. B210910
Mecarbinate (CAS 15574-49-9) is a small-molecule inhibitor targeting hepatitis C virus (HCV) replication It is designed to inhibit viral replication thereby interfering with essential HCV biological processes Mecarbinate exerts its biological activity primarily through disruption of viral RNA replication potentially by inhibiting viral RNA polymerase or host factors necessary for genome replication In in vitro assays Mecarbinate demonstrates inhibition of HCV RNA replication efficiency with reported IC50 values varying according to experimental context and assay specifics Based on these pharmacological properties Mecarbinate holds research potential in studying viral infection mechanisms and aiding antiviral therapy screening