Learn More
Andwin Scientific CryoPro Storage Boxes Mechanical Cryogenic Freezer Boxes without divide 12.7 x 12.7 x 7.6 cm
SDP
Catalog No. NC9370130
Encompass_Preferred
CryoPro Storage Boxes Mechanical Cryogenic Freezer Boxes without divider. Constructed of durable fiberboard with a protective moisture-repellant coating. Dimensions: 12.7 x 12.7 x 7.6 cm. Do not feature slots and cannot be used with LN2., 1/PK
Catalog No. NC9370130 Supplier Andwin Scientific Supplier No. 82021114
CryoPro Storage Boxes Mechanical Cryogenic Freezer Boxes without divider. Constructed of durable fiberboard with a protective moisture-repellant coating. Dimensions: 12.7 x 12.7 x 7.6 cm. Do not feature slots and cannot be used with LN2., 1/PK