Learn More
Andwin Scientific MECH CRYO FRZR BX 5X5X2
SDP
Catalog No. NC0345438
Encompass_Preferred
Mechanical Cryogenic Freezer Box No divider. NO drain slots - Not for Use with LN(2). Dimensions:5 x 5 x 2 inches. Constructed of durable fiberboard with a protective moisture repellant coating.
Catalog No. NC0345438 Supplier Andwin Scientific Supplier No. 82007142
Mechanical Cryogenic Freezer Box No divider. NO drain slots - Not for Use with LN(2). Dimensions:5 x 5 x 2 inches. Constructed of durable fiberboard with a protective moisture repellant coating.