Learn More
Grainger Mech Expansion Plug Hex Nut 2.3 H 2 Pipe
Catalog No. 502751243
Encompass_Preferred
The Grainger 14M038 is a 2-3/8" hex nut expansion plug for 2-3/8" pipe. It is made from malleable iron and has a working pressure of 300 psi. This plug features a tapered thread design for easy installation and removal. It can be used to seal open ends of pipe or close off unused outlets. The hex nut allows for wrench tightening during installation. This is an ideal plug for temporarily sealing pipe openings during maintenance or construction.
Catalog No. 50-275-1243 Supplier Grainger Supplier No. 14M038
The Grainger 14M038 is a 2-3/8" hex nut expansion plug for 2-3/8" pipe. It is made from malleable iron and has a working pressure of 300 psi. This plug features a tapered thread design for easy installation and removal. It can be used to seal open ends of pipe or close off unused outlets. The hex nut allows for wrench tightening during installation. This is an ideal plug for temporarily sealing pipe openings during maintenance or construction.