Learn More
Grainger Mech Expansion Plug Hex Nut 2.3 H
Catalog No. 502752640
Encompass_Preferred
The Shaw Plugs 14M036 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in hardened concrete and can withstand up to 75 lbs of tension load. The anchor body is knurled for better gripping and requires a 3/8" diameter hole to be predrilled. It expands upon installation with a standard socket, hammer, or impact wrench. This is a reusable anchor that can be easily removed and reinstalled if needed. The anchor is made in the USA and comes in a package of 25 pieces.
Catalog No. 50-275-2640 Supplier Grainger Supplier No. 14M036
The Shaw Plugs 14M036 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in hardened concrete and can withstand up to 75 lbs of tension load. The anchor body is knurled for better gripping and requires a 3/8" diameter hole to be predrilled. It expands upon installation with a standard socket, hammer, or impact wrench. This is a reusable anchor that can be easily removed and reinstalled if needed. The anchor is made in the USA and comes in a package of 25 pieces.