Learn More
Grainger Mech Expansion Plug Hex Nut 2.3 H
Catalog No. 502752015
Encompass_Preferred
The Shaw Plugs 14M037 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in cracked and uncracked normal-weight concrete and can be installed in drilled holes to provide an anchoring point. The expansion plug features a cone expander with zinc plating for corrosion resistance. It is rated for tension loads up to 60 lbs and shear loads up to 45 lbs in 2,500 psi concrete when installed at the full embedment depth.
Catalog No. 50-275-2015 Supplier Grainger Supplier No. 14M037
The Shaw Plugs 14M037 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in cracked and uncracked normal-weight concrete and can be installed in drilled holes to provide an anchoring point. The expansion plug features a cone expander with zinc plating for corrosion resistance. It is rated for tension loads up to 60 lbs and shear loads up to 45 lbs in 2,500 psi concrete when installed at the full embedment depth.