Learn More
Grainger Mech Expansion Plug Snap Handle 2.53 H
Catalog No. 502752404
Encompass_Preferred
The Grainger 14M022 is a Westward 3/8-inch drive deep impact socket set made in Taiwan. This 25-piece socket set includes sizes ranging from 4mm to 14mm, as well as a 3-inch extension, universal joint, and storage case. The chrome vanadium steel sockets feature a black phosphate finish for corrosion resistance. This impact socket set is designed for use with impact wrenches in heavy-duty applications. The blow-molded case provides convenient storage and portability.
Catalog No. 50-275-2404 Supplier Grainger Supplier No. 14M022
The Grainger 14M022 is a Westward 3/8-inch drive deep impact socket set made in Taiwan. This 25-piece socket set includes sizes ranging from 4mm to 14mm, as well as a 3-inch extension, universal joint, and storage case. The chrome vanadium steel sockets feature a black phosphate finish for corrosion resistance. This impact socket set is designed for use with impact wrenches in heavy-duty applications. The blow-molded case provides convenient storage and portability.