Learn More
Grainger Mech Expansion Plug Snap Handle 2.62 H
Catalog No. 502752435
Encompass_Preferred
The Shaw Plugs 14M015 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in cracked and uncracked normal-weight concrete and can be installed in drilled holes to provide an anchoring point. The expansion plug features a cone expander with zinc plating for corrosion resistance. It is rated for tension loads up to 60 lbs and shear loads up to 45 lbs in 2,500 psi concrete when installed at the full embedment depth.
Catalog No. 50-275-2435 Supplier Grainger Supplier No. 14M015
The Shaw Plugs 14M015 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in cracked and uncracked normal-weight concrete and can be installed in drilled holes to provide an anchoring point. The expansion plug features a cone expander with zinc plating for corrosion resistance. It is rated for tension loads up to 60 lbs and shear loads up to 45 lbs in 2,500 psi concrete when installed at the full embedment depth.