Learn More
Grainger Mech Expansion Plug Snap Handle 2.53 H
Catalog No. 502748671
Encompass_Preferred
The MECH Expansion Plug with Snap Handle from Grainger is a 2-53"H mechanical expansion plug made of zinc with a snap handle for setting. It has a 5/8" nominal size and is designed for use in concrete with a temperature range of -65 to 300°F. This zinc expansion plug provides a tight, vibration-resistant fit and is ideal for commercial and industrial applications. The snap handle allows for quick and easy setting.
Catalog No. 50-274-8671 Supplier Grainger Supplier No. 14M017
The MECH Expansion Plug with Snap Handle from Grainger is a 2-53"H mechanical expansion plug made of zinc with a snap handle for setting. It has a 5/8" nominal size and is designed for use in concrete with a temperature range of -65 to 300°F. This zinc expansion plug provides a tight, vibration-resistant fit and is ideal for commercial and industrial applications. The snap handle allows for quick and easy setting.