Learn More
Grainger Mech Expansion Plug T-Handle 1.95 H
Catalog No. 502753041
Encompass_Preferred
The Shaw Plugs 14M004 Mechanical Expansion Plug is a 3/8" diameter zinc-plated steel mechanical expansion anchor with a 1-1/4" embedment depth. It is designed for use in concrete and can withstand up to 75 lbs of load. The anchor has a split end that expands outward when a screw is inserted to grip the sides of the hole. This is an economical basic anchor for light duty applications.
Catalog No. 50-275-3041 Supplier Grainger Supplier No. 14M004
The Shaw Plugs 14M004 Mechanical Expansion Plug is a 3/8" diameter zinc-plated steel mechanical expansion anchor with a 1-1/4" embedment depth. It is designed for use in concrete and can withstand up to 75 lbs of load. The anchor has a split end that expands outward when a screw is inserted to grip the sides of the hole. This is an economical basic anchor for light duty applications.