Learn More
Grainger Mech Expansion Plug T-Handle 2.07 H
Catalog No. 502753039
Encompass_Preferred
The Shaw Plugs Mechanical Expansion Plug is a mechanical expansion anchor made of zinc plated steel with a 3/8" diameter and 1-3/4" length. It has a hex head and is designed for use in hardened concrete. The anchor is inserted into a pre-drilled hole and then tightened to expand the anchor for a secure hold. This product can be used for hanging pipe, conduit, light fixtures, signs, and other applications that require a secure mounting point in concrete. The anchor has a temperature range of -20°F to 150°F and meets ASTM F593 standards. It is sold individually.
Catalog No. 50-275-3039 Supplier Grainger Supplier No. 14M013
The Shaw Plugs Mechanical Expansion Plug is a mechanical expansion anchor made of zinc plated steel with a 3/8" diameter and 1-3/4" length. It has a hex head and is designed for use in hardened concrete. The anchor is inserted into a pre-drilled hole and then tightened to expand the anchor for a secure hold. This product can be used for hanging pipe, conduit, light fixtures, signs, and other applications that require a secure mounting point in concrete. The anchor has a temperature range of -20°F to 150°F and meets ASTM F593 standards. It is sold individually.