Learn More
Grainger Mech Expansion Plug T-Handle 2.55 H
Catalog No. 502753042
Encompass_Preferred
The Shaw Plugs 14M006 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in cracked and uncracked normal-weight concrete and can be installed in drilled holes to provide an anchoring point. The expansion plug features a cone expander with zinc plating for corrosion resistance. It is rated for tension loads up to 60 lbs and shear loads up to 45 lbs in 2,500 psi concrete when installed at the full embedment depth.
Catalog No. 50-275-3042 Supplier Grainger Supplier No. 14M006
The Shaw Plugs 14M006 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in cracked and uncracked normal-weight concrete and can be installed in drilled holes to provide an anchoring point. The expansion plug features a cone expander with zinc plating for corrosion resistance. It is rated for tension loads up to 60 lbs and shear loads up to 45 lbs in 2,500 psi concrete when installed at the full embedment depth.