Learn More
Grainger Mech Expansion Plug T-Handle 3.86 H
Catalog No. 502748753
Encompass_Preferred
The Shaw Plugs 14M040 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in concrete and can support up to 75 lbs in tension and 225 lbs in shear. The anchor body has a cone shape and it expands outward when the screw is tightened to grip the sides of the hole. This is a mechanical type anchor that does not require setting with a hammer. It is easy to install by pre-drilling a hole, inserting the anchor, and tightening the screw to expand and set the anchor in place. The anchor is made in the USA and is ETL listed to US safety standards.
Catalog No. 50-274-8753 Supplier Grainger Supplier No. 14M040
The Shaw Plugs 14M040 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in concrete and can support up to 75 lbs in tension and 225 lbs in shear. The anchor body has a cone shape and it expands outward when the screw is tightened to grip the sides of the hole. This is a mechanical type anchor that does not require setting with a hammer. It is easy to install by pre-drilling a hole, inserting the anchor, and tightening the screw to expand and set the anchor in place. The anchor is made in the USA and is ETL listed to US safety standards.