Learn More
Grainger Mech Expansion Plug T-Handle 3.97 H
Catalog No. 502752190
Encompass_Preferred
The Shaw Plugs 14M011 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in hardened concrete and can withstand tension loads up to 50 lbs and shear loads up to 30 lbs. The expansion plug features a split expansion clip and cone expander for easy installation by hammering directly into a pre-drilled hole. Once set, it provides a permanent threaded anchor point to attach fixtures and other hardware to concrete surfaces. This zinc alloy plug offers corrosion resistance for long-lasting durability.
Catalog No. 50-275-2190 Supplier Grainger Supplier No. 14M011
The Shaw Plugs 14M011 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in hardened concrete and can withstand tension loads up to 50 lbs and shear loads up to 30 lbs. The expansion plug features a split expansion clip and cone expander for easy installation by hammering directly into a pre-drilled hole. Once set, it provides a permanent threaded anchor point to attach fixtures and other hardware to concrete surfaces. This zinc alloy plug offers corrosion resistance for long-lasting durability.