Learn More
Grainger Mech Expansion Plug T-Handle 3.87 H
Catalog No. 502752432
Encompass_Preferred
The Grainger 14M012 is a 14-inch diameter, 12-inch long, cast iron pulley with a 1-1/8 inch bore and a 1/2 inch keyway. It has an A-style hub, is rated for 1750 RPM max, and can transmit up to 15 HP. This pulley weighs approximately 35 pounds and is made in the USA. It can be used with v-belts and has a rough finish for good belt grip. The max operating temperature is 300°F.
Catalog No. 50-275-2432 Supplier Grainger Supplier No. 14M012
The Grainger 14M012 is a 14-inch diameter, 12-inch long, cast iron pulley with a 1-1/8 inch bore and a 1/2 inch keyway. It has an A-style hub, is rated for 1750 RPM max, and can transmit up to 15 HP. This pulley weighs approximately 35 pounds and is made in the USA. It can be used with v-belts and has a rough finish for good belt grip. The max operating temperature is 300°F.