Learn More
Grainger Mech Expansion Plug T-Handle 3.87 H
Catalog No. 502752626
Encompass_Preferred
The Shaw Plugs 14M042 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in concrete and can support up to 75 lbs in tension and 225 lbs in shear. The expansion plug has a zinc finish and includes a 3/8" diameter screw for fastening. Overall it provides a secure and reusable anchoring point in concrete walls and ceilings.
Catalog No. 50-275-2626 Supplier Grainger Supplier No. 14M042
The Shaw Plugs 14M042 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in concrete and can support up to 75 lbs in tension and 225 lbs in shear. The expansion plug has a zinc finish and includes a 3/8" diameter screw for fastening. Overall it provides a secure and reusable anchoring point in concrete walls and ceilings.