Learn More
Grainger Mech Expansion Plug Wing Nut 1.69 H
Catalog No. 502752446
Encompass_Preferred
The Shaw Plugs 14M028 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in cracked and uncracked normal-weight concrete and can be installed in drilled holes to provide an anchoring point. The expansion plug features a cone expander and dome head for fixing lightweight loads. It meets ASTM F593 standards and offers corrosion resistance. With a temperature range of -20°F to 180°F, this mechanical anchor is well-suited for indoor and outdoor applications. The expansion plug comes in a package of 25 pieces.
Catalog No. 50-275-2446 Supplier Grainger Supplier No. 14M028
The Shaw Plugs 14M028 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in cracked and uncracked normal-weight concrete and can be installed in drilled holes to provide an anchoring point. The expansion plug features a cone expander and dome head for fixing lightweight loads. It meets ASTM F593 standards and offers corrosion resistance. With a temperature range of -20°F to 180°F, this mechanical anchor is well-suited for indoor and outdoor applications. The expansion plug comes in a package of 25 pieces.