Learn More
Grainger Mech Expansion Plug Wing Nut 2.3 H
Catalog No. 502751210
Encompass_Preferred
The Shaw Plugs 14M031 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in hardened concrete and can support up to 75 lbs in tension and 225 lbs in shear. The expansion plug has a split end that expands outward when a screw is inserted to provide holding power. It requires a 3/8" diameter hole to be predrilled before insertion. This is an economical zinc alloy anchor ideal for lightweight duty in concrete.
Catalog No. 50-275-1210 Supplier Grainger Supplier No. 14M031
The Shaw Plugs 14M031 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in hardened concrete and can support up to 75 lbs in tension and 225 lbs in shear. The expansion plug has a split end that expands outward when a screw is inserted to provide holding power. It requires a 3/8" diameter hole to be predrilled before insertion. This is an economical zinc alloy anchor ideal for lightweight duty in concrete.