Learn More
BRANSON ULTRASONICS CORPORATION (3P) MECH. HEATER ULTRASONIC BATH
Catalog No. NC3721465
Encompass_Preferred
Branson Ultrasonics MH Series Heated Ultrasonic Cleaning Bath, Model: M5800H, Capacity: 9.5L (2.5 gal.), Dimensions Interior: 29 x 24 x 15cm (11.5 x 9.5 x 6 in.), Voltage: 120V, Includes: Tank cover and drain, Plastic, Timer: 60 min., Warranty: 2 Years, Weight: 14 lb., 6.4kg
Catalog No. NC3721465 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPX952517R
Branson Ultrasonics MH Series Heated Ultrasonic Cleaning Bath, Model: M5800H, Capacity: 9.5L (2.5 gal.), Dimensions Interior: 29 x 24 x 15cm (11.5 x 9.5 x 6 in.), Voltage: 120V, Includes: Tank cover and drain, Plastic, Timer: 60 min., Warranty: 2 Years, Weight: 14 lb., 6.4kg