Learn More
BRANSON ULTRASONICS CORPORATION (3P) MECH. HEATER ULTRASONIC BATH
Supplier:  BRANSON ULTRASONICS CORPORATION (3P) CPX952817R
Encompass_Preferred
Branson Ultrasonics MH Series Heated Ultrasonic Cleaning Bath, Model: M8800H, Capacity: 20.8L (5.5 gal.), Dimensions Interior: 49.5 x 29 x 15cm (19.5 x 11.5 x 6 in.), Voltage: 120V, Includes: Tank cover and drain, Plastic, Timer: 60 min., Warranty: 2 Years, Weight: 26 lb., 11.8kg
Catalog No. NC3721468