Learn More
BRANSON ULTRASONICS CORPORATION (3P) MECH. HEATER ULTRASONIC BATH
Catalog No. NC3721469
Encompass_Preferred
Branson Ultrasonics MH Series Heated Ultrasonic Cleaning Bath, Model: M1800H-E, Capacity: 1.9L (0.5 gal.), Dimensions Interior: 15 x 14 x 10cm (6 x 5.5 x 4 in.), Voltage: 230/240V, Includes: Tank cover, Plastic, Timer: 60 min., Warranty: 2 Years, Weight: 7 lb., 3.2kg
Catalog No. NC3721469 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPX952137R
Branson Ultrasonics MH Series Heated Ultrasonic Cleaning Bath, Model: M1800H-E, Capacity: 1.9L (0.5 gal.), Dimensions Interior: 15 x 14 x 10cm (6 x 5.5 x 4 in.), Voltage: 230/240V, Includes: Tank cover, Plastic, Timer: 60 min., Warranty: 2 Years, Weight: 7 lb., 3.2kg