Learn More
BRANSON ULTRASONICS CORPORATION (3P) MECH. HEATER ULTRASONIC BATH
Catalog No. NC3721463
Encompass_Preferred
Branson Ultrasonics MH Series Heated Ultrasonic Cleaning Bath, Model: M2800H, Capacity: 2.8L (0.75 gal.), Dimensions Interior: 23 x 14 x 10cm (9.5 x 5.5 x 4 in.), Voltage: 120V, Includes: Tank cover, Plastic, Timer: 60 min., Warranty: 2 Years, Weight: 9 lb., 4.1kg
Catalog No. NC3721463 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPX952217R
Branson Ultrasonics MH Series Heated Ultrasonic Cleaning Bath, Model: M2800H, Capacity: 2.8L (0.75 gal.), Dimensions Interior: 23 x 14 x 10cm (9.5 x 5.5 x 4 in.), Voltage: 120V, Includes: Tank cover, Plastic, Timer: 60 min., Warranty: 2 Years, Weight: 9 lb., 4.1kg