Learn More
BRANSON ULTRASONICS CORPORATION (3P) MECH. HEATER ULTRASONIC BATH
Catalog No. NC3721464
Encompass_Preferred
Branson Ultrasonics MH Series Heated Ultrasonic Cleaning Bath, Model: M3800H, Capacity: 5.7L (1.5 gal.), Dimensions Interior: 29 x 15 x 15cm (11.5 x 6 x 6 in.), Voltage: 120V, Includes: Tank cover and drain, Plastic, Timer: 60 min., Warranty: 2 Years, Weight: 12 lb., 5.4kg
Catalog No. NC3721464 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPX952317R
Branson Ultrasonics MH Series Heated Ultrasonic Cleaning Bath, Model: M3800H, Capacity: 5.7L (1.5 gal.), Dimensions Interior: 29 x 15 x 15cm (11.5 x 6 x 6 in.), Voltage: 120V, Includes: Tank cover and drain, Plastic, Timer: 60 min., Warranty: 2 Years, Weight: 12 lb., 5.4kg