Learn More
Rice Lake Weighing System MECH SCALE LB & KG W/ HT ROD
Catalog No. 50597129
Encompass_Preferred
Medical Scale, Rice Lake Weighing Systems, RL-MPS Mechanical Physician Scale 440lb(200kg) x 4oz (100g) including retractable height rod and rear wheels, 10.75in x 14.75in Platform, Easy operation w/ two sliding weights
Catalog No. 50-597-129 Supplier Rice Lake Weighing System Supplier No. 102613
Medical Scale, Rice Lake Weighing Systems, RL-MPS Mechanical Physician Scale 440lb(200kg) x 4oz (100g) including retractable height rod and rear wheels, 10.75in x 14.75in Platform, Easy operation w/ two sliding weights