Learn More
MSC Supco Mechanical Wall Thermostat, Heat and Cool, 24 VAC, Horizontal, 1 ea.
Catalog No. 5000507718
Encompass
This single-stage mechanical wall thermostat is designed for precise temperature control in heating and cooling systems. It is compatible with most 4 or 5 wire 24 VAC systems and single-stage heat pumps, providing reliable performance for various HVAC applications.
  • Features a horizontal mounting configuration
  • Includes a manual changeover switch
  • Equipped with an adjustable heat anticipator
  • Operates via battery power with two included AA batteries
  • Includes a low battery indicator and easy front-load access
  • Features a soft touch keypad and blue back light for enhanced usability
  • Provides quiet switching functionality and latching relays
  • Mercury free design
  • Integrated thermometer with Fahrenheit scale
Catalog No. 50-005-07718 Supplier MSC Supplier No. 91965251
This single-stage mechanical wall thermostat is designed for precise temperature control in heating and cooling systems. It is compatible with most 4 or 5 wire 24 VAC systems and single-stage heat pumps, providing reliable performance for various HVAC applications.
  • Features a horizontal mounting configuration
  • Includes a manual changeover switch
  • Equipped with an adjustable heat anticipator
  • Operates via battery power with two included AA batteries
  • Includes a low battery indicator and easy front-load access
  • Features a soft touch keypad and blue back light for enhanced usability
  • Provides quiet switching functionality and latching relays
  • Mercury free design
  • Integrated thermometer with Fahrenheit scale