Learn More
Ace Glass, Inc. Bearing, PTFE, non-shedding, low-vacuum, 24/40 inner joint, accepts 10mm OD stir shaft
SDP
Catalog No. NC0333148
Encompass
Bearing, PTFE, non-shedding, low-vacuum, 24/40 inner joint, accepts 10mm OD stir shaft
Catalog No. NC0333148 Supplier Ace Glass, Inc. Supplier No. 805002
Bearing, PTFE, non-shedding, low-vacuum, 24/40 inner joint, accepts 10mm OD stir shaft