Learn More
Andwin Scientific 2" Mechanical Cryogenic box, no drain slots, with grids, 81 cell divider
SDP
Catalog No. NC1776394
Encompass_Preferred
VWR Mechanical Cryogenic Freezer Box 81 cell divider. NO drain slots - Not for Use with LN(2). Dimensions:5 x 5 x 2 inches. Constructed of durable fiberboard with a protective moisture repellant coating. Numbered grid printed on lid.
Catalog No. NC1776394 Supplier Andwin Scientific Supplier No. SB2CC811P
VWR Mechanical Cryogenic Freezer Box 81 cell divider. NO drain slots - Not for Use with LN(2). Dimensions:5 x 5 x 2 inches. Constructed of durable fiberboard with a protective moisture repellant coating. Numbered grid printed on lid.