Learn More
Andwin Scientific MECHANICAL CRYOGENIC BOX EACH
SDP
Catalog No. NC1765082
Encompass_Preferred
VWR 4" Mechanical Cryogenic box, no drain slots, no grids 13.2 x 13.2 x 10.2 cm (51/5 x 51/5 x 4"), including lid
Catalog No. NC1765082 Supplier Andwin Scientific Supplier No. 89214776
VWR 4" Mechanical Cryogenic box, no drain slots, no grids 13.2 x 13.2 x 10.2 cm (51/5 x 51/5 x 4"), including lid