Learn More
Electron Microscopy Sciences Mechanical Tool Kit
SDP
Catalog No. 501901607
Encompass
The mechanical sample preparation prior to thinning requires custom made mechanical hand tools. Containing six handy, reliable pieces, this kit has been designed on the basis of practical experience of many users. Some of the tools are suitable for adjusting the sample pieces into the Ti discs, others for removing the excess glue and cutting, or adjusting the seat plate of the sample holder.
Catalog No. 50-190-1607 Supplier Electron Microscopy Sciences Supplier No. 55110HSS
The mechanical sample preparation prior to thinning requires custom made mechanical hand tools. Containing six handy, reliable pieces, this kit has been designed on the basis of practical experience of many users. Some of the tools are suitable for adjusting the sample pieces into the Ti discs, others for removing the excess glue and cutting, or adjusting the seat plate of the sample holder.