Learn More
BRANSON ULTRASONICS CORPORATION (3P) MECHANICAL ULTRASONIC BATHS
Catalog No. NC3721438
Encompass_Preferred
Branson Ultrasonics M Series Ultrasonic Cleaning Bath, Model: M2800, Voltage: 120V, Capacity: 2.8L (0.75 gal.), Dimensions Interior: 23 x 14 x 10cm (9.5 x 5.5 x 4 in.), Hertz: 40kHz, Includes: Tank cover, mechanical timer, Plastic Housing, Stainless Steel Tank, Timer: 60 min., Warranty: 2 Years
Catalog No. NC3721438 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPX952216R
Branson Ultrasonics M Series Ultrasonic Cleaning Bath, Model: M2800, Voltage: 120V, Capacity: 2.8L (0.75 gal.), Dimensions Interior: 23 x 14 x 10cm (9.5 x 5.5 x 4 in.), Hertz: 40kHz, Includes: Tank cover, mechanical timer, Plastic Housing, Stainless Steel Tank, Timer: 60 min., Warranty: 2 Years