Learn More
BRANSON ULTRASONICS CORPORATION (3P) MECHANICAL ULTRASONIC BATHS
Supplier:  BRANSON ULTRASONICS CORPORATION (3P) CPX952316R
Encompass_Preferred
Branson Ultrasonics M Series Ultrasonic Cleaning Bath, Model: M3800, Voltage: 120V, Capacity: 5.7L (1.5 gal.), Dimensions Interior: 29 x 15 x 15cm (11.5 x 6 x 6 in.), Hertz: 40kHz, Includes: Tank cover, Length Tank: 29cm, Plastic Housing, Stainless Steel Tank, Timer: 60 min., Warranty: 2 Years
Catalog No. NC3721440